Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146314.1 | NP_001139786.1 | 4 | 1204 | ||
NM_001254753.1 | NP_001241682.1 | 3 | 965 | |||
NM_032750.2 | NP_116139.1 | 4 | 1398 | |||
GenBank mRNA | AK075112.1 | - | 4 | 789 | ||
AK092152.1 | - | 1 | 1589 | |||
AK096073.1 | - | 3 | 965 | |||
AK296599.1 | - | 4 | 459 | |||
AK298022.1 | - | 5 | 537 | |||
AK298813.1 | - | 3 | 932 | |||
BC007234.1 | - | 4 | 782 | 2823284 | ||
BC050650.1 | - | 4 | 596 | |||
BC056411.1 | - | 4 | 605 | 6048106 | ||
BC071931.1 | - | 5 | 1017 | |||
FJ224306.1 | - | 625 |