Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193308.1 | NP_001180237.1 | 3 | 499 | ||
NM_032872.2 | NP_116261.1 | 3 | 499 | |||
XM_005246022.3 | XP_005246079.1 | 3 | 654 | |||
XM_006710990.1 | XP_006711053.1 | 3 | 654 | |||
XM_006710991.1 | XP_006711054.1 | 3 | 450 | |||
GenBank mRNA | AK027902.1 | - | 3 | 469 | ||
AK074154.1 | - | 2 | 477 | |||
AK096437.1 | - | 3 | 651 | |||
AK293038.1 | - | 2 | 765 | |||
AY037157.1 | - | 3 | 426 | |||
BC009224.2 | - | 1 | 30 | |||
BC015764.1 | - | 2 | 242 | 4851468 | ||
BC035725.1 | - | 3 | 406 | 5722683 | ||
DA435960.1 | - | 3 | 499 | |||
DA948740.1 | - | 3 | 476 |