Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182491.2 | NP_872297.2 | 5 | 657 | ||
XM_017012774.1 | XP_016868263.1 | 5 | 657 | |||
GenBank mRNA | AI435373.1 | - | 1 | 247 | ||
AK290170.1 | - | 5 | 616 | |||
BC016963.1 | - | 5 | 609 | |||
BC029558.1 | - | 5 | 600 | 5263788 |