Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033513.2 | NP_277048.2 | 2 | 823 | ||
GenBank mRNA | AK126170.1 | - | 3 | 766 | ||
BC009520.1 | - | 2 | 709 |