Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017971.2 | NP_001017971.1 | 7, 8 | 1399 | ||
XM_011543723.2 | XP_011542025.1 | 7, 8 | 2046 | |||
XM_017010039.1 | XP_016865528.1 | 2, 3 | 205 | |||
XM_017010040.1 | XP_016865529.1 | 2, 3 | 110 | |||
XM_017010041.1 | XP_016865530.1 | 8, 9 | 2124 | |||
XM_017010042.1 | XP_016865531.1 | 9, 10 | 2030 | |||
XM_017010043.1 | XP_016865532.1 | 8, 9 | 2113 | |||
XM_017010044.1 | XP_016865533.1 | 7, 8 | 2072 | |||
XM_017010045.1 | XP_016865534.1 | 7, 8 | 1624 | |||
XM_017010046.1 | XP_016865535.1 | 7, 8 | 1705 | |||
XM_017010047.1 | XP_016865536.1 | 6, 7 | 1994 | |||
GenBank mRNA | AK022625.1 | - | 7, 8 | 1399 | ||
BC093982.1 | - | 1, 2 | 129 | 7939827 | ||
BC112191.1 | - | 1, 2 | 129 | 8327659 |