Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291726.1 | NP_001278655.1 | 5, 6 | 592 | ||
NM_001291728.1 | NP_001278657.1 | 5, 6 | 682 | |||
NM_033419.4 | NP_219487.3 | 6, 7 | 745 | |||
XM_011525481.2 | XP_011523783.1 | 7, 8 | 874 | |||
GenBank mRNA | AB088396.1 | - | 6, 7 | 697 | ||
AK075474.1 | - | 5, 6 | 579 | |||
AK294639.1 | - | 5, 6 | 669 | |||
AK301106.1 | - | 7, 8 | 647 | |||
AK314469.1 | - | 6, 7 | 705 | |||
AY358437.1 | - | 6, 7 | 721 | |||
BC010652.1 | - | 6, 7 | 706 | 3855206 |