Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052872.3 | NP_443104.1 | 2 | 275 | ||
XM_011514276.1 | XP_011512578.1 | 3 | 341 | |||
GenBank mRNA | AF332389.1 | - | 1 | 388 | ||
AF384857.1 | - | 2 | 204 | |||
BC070124.1 | - | 2 | 275 | 30417554 | ||
JF305974.1 | - | 2 | 204 |