Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001653.2 | NP_001001653.1 | 2 | 131 | ||
NM_052877.4 | NP_443109.2 | 2 | 70 | |||
NM_201542.4 | NP_963836.2 | 2 | 70 | |||
GenBank mRNA | AF521562.1 | - | 1 | 22 | ||
AV684532.1 | - | 2 | 107 | |||
BC010019.2 | - | 2 | 103 | 2905623 | ||
BC010543.1 | - | 11 | ||||
BG620658.1 | - | 2 | 71 | |||
BI488866.1 | - | 1 | 26 |