Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099678.1 | NP_001093148.1 | 1, 2 | 601 | ||
GenBank mRNA | BC013757.1 | - | 1, 2 | 601 |