Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058173.2 | NP_477521.1 | 2, 3 | 171 | ||
GenBank mRNA | AF414087.1 | - | 2, 3 | 149 | ||
AI905624.1 | - | 2, 3 | 172 | |||
AY359062.1 | - | 2, 3 | 142 | |||
AY651259.1 | - | 2, 3 | 103 | |||
BC111421.1 | - | 2, 3 | 136 | 40021582 |