Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFYVE27 SIRNA Assays ›
Gene Symbol:
ZFYVE27
Gene Name:
zinc finger FYVE-type containing 27
Gene Aliases:
PROTRUDIN, SPG33
Chromosome Location:
Chr.10: 97737121 - 97760907 on Build GRCh38
UniGene ID:
Hs.744075
Species:
Human
siRNA ID
HSS151127
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFYVE27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002261.3
NP_001002261.1
6
731
NM_001002262.3
NP_001002262.1
7
936
NM_001174119.1
NP_001167590.1
6
836
NM_001174120.1
NP_001167591.1
5
674
NM_001174121.1
NP_001167592.1
5
663
NM_001174122.1
NP_001167593.1
4
578
NM_144588.6
NP_653189.3
7
936
XM_005269502.3
XP_005269559.1
7
925
XM_005269503.3
XP_005269560.1
7
929
XM_005269504.3
XP_005269561.1
7
925
XM_005269505.4
XP_005269562.1
7
925
XM_005269506.3
XP_005269563.1
7
925
XM_005269508.3
XP_005269565.1
6
854
XM_005269509.3
XP_005269566.1
6
858
XM_005269510.3
XP_005269567.1
5
670
XM_005269511.3
XP_005269568.1
5
676
XM_011539252.2
XP_011537554.1
7
925
XM_011539253.2
XP_011537555.1
7
925
XM_011539254.2
XP_011537556.1
7
900
XM_011539255.2
XP_011537557.1
7
907
XM_017015644.1
XP_016871133.1
7
925
XM_017015645.1
XP_016871134.1
7
925
XM_017015646.1
XP_016871135.1
7
925
XM_017015647.1
XP_016871136.1
7
929
XM_017015648.1
XP_016871137.1
5
670
XM_017015649.1
XP_016871138.1
5
670
XM_017015650.1
XP_016871139.1
7
900
XM_017015651.1
XP_016871140.1
6
854
XM_017015652.1
XP_016871141.1
6
858
XM_017015653.1
XP_016871142.1
6
854
XM_017015654.1
XP_016871143.1
6
858
XM_017015655.1
XP_016871144.1
5
676
GenBank mRNA
AK057481.1
-
7
907
AK074876.1
-
7
917
AK097945.1
-
5
667
AK296295.1
-
5
663
AK296588.1
-
6
781
AK299735.1
-
4
571
AK300438.1
-
7
929
AL834235.1
-
6
665
BC030621.2
-
7
929
4818199
BU838354.1
-
3
254
BX647836.1
-
7
871
KU178847.1
-
5
526
KU178848.1
-
5
526
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
neuron projection development
neurotrophin TRK receptor signaling pathway
protein localization to plasma membrane
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0