Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017923.1 | NP_001017923.1 | 3, 4 | 886 | ||
GenBank mRNA | AF250395.1 | - | 3, 4 | 870 | ||
AK126985.1 | - | 2, 3 | 642 | |||
BC030258.1 | - | 2, 3 | 624 | |||
BC105030.1 | - | 3, 4 | 879 | 8144033 | ||
BC105032.1 | - | 3, 4 | 879 | 8144035 | ||
BU624091.1 | - | 2, 3 | 290 |