Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317225.1 | NP_001304154.1 | 4 | 663 | ||
NM_001317226.1 | NP_001304155.1 | 3 | 286 | |||
NM_173847.4 | NP_776246.1 | 4 | 595 | |||
GenBank mRNA | AF216311.1 | - | 4 | 595 | ||
AY358653.1 | - | 5 | 722 | |||
BC029867.1 | - | 4 | 663 | 5169651 | ||
BC100886.2 | - | 4 | 553 | 40003360 | ||
BC100887.1 | - | 4 | 553 | 40003365 | ||
BF511575.1 | - | 2 | 239 | |||
BQ438507.1 | - | 3 | 281 |