Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF391162.1 | - | 3 | 303 | ||
AF391163.1 | - | 4 | 308 | |||
AF391164.1 | - | 2 | 301 | |||
AK130199.1 | - | 4 | 332 |