Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002901.3 | NP_001002901.1 | 5 | 360 | ||
NM_001288829.1 | NP_001275758.1 | 3 | 146 | |||
NM_001288830.1 | NP_001275759.1 | 2 | 125 | |||
NM_001288831.1 | NP_001275760.1 | 3 | 146 | |||
NM_001288832.1 | NP_001275761.1 | 2 | 125 | |||
NM_001320241.1 | NP_001307170.1 | 4 | 174 | |||
GenBank mRNA | AK290733.1 | - | 4 | 174 | ||
AY670683.1 | - | 3 | 141 | |||
AY670684.1 | - | 3 | 141 | |||
AY670685.1 | - | 2 | 120 | |||
AY670686.1 | - | 3 | 141 | |||
AY670687.1 | - | 2 | 120 | |||
AY786314.1 | - | 3 | 126 | |||
BX364259.2 | - | 4 | 320 | |||
DQ021957.1 | - | 5 | 360 |