Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138797.2 | NP_620152.1 | 6 | 810 | ||
NR_036556.1 | 6 | 805 | ||||
XM_006724136.1 | XP_006724199.1 | 5 | 689 | |||
XM_006724137.1 | XP_006724200.1 | 5 | 750 | |||
XM_011529877.2 | XP_011528179.1 | 6 | 1233 | |||
XM_011529878.2 | XP_011528180.1 | 6 | 1272 | |||
XM_011529879.2 | XP_011528181.1 | 7 | 660 | |||
XM_011529880.2 | XP_011528182.1 | 5 | 665 | |||
XM_011529882.2 | XP_011528184.1 | 6 | 592 | |||
XM_011529883.2 | XP_011528185.1 | 6 | 823 | |||
XM_011529884.2 | XP_011528186.1 | 6 | 625 | |||
XM_017028590.1 | XP_016884079.1 | 6 | 1140 | |||
XM_017028591.1 | XP_016884080.1 | 6 | 588 | |||
XM_017028593.1 | XP_016884082.1 | 6 | 1075 | |||
GenBank mRNA | AK127583.1 | - | 3 | 1120 | ||
BC014641.1 | - | 6 | 760 | 2988073 | ||
BC066909.1 | - | 6 | 777 | 4795922 | ||
BX401122.2 | - | 6 | 808 | |||
CR456471.1 | - | 6 | 770 |