Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009609.3 | NP_001009609.2 | 2 | 317 | ||
XM_017029264.1 | XP_016884753.1 | 2 | 437 | |||
GenBank mRNA | BC062750.1 | - | 321 | 6618011 | ||
DQ336128.1 | - | 2 | 233 | |||
DQ336129.1 | - | 2 | 233 |