Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039582.3 | NP_001034671.3 | 2 | 470 | ||
NM_001135740.1 | NP_001129212.1 | 2 | 104 | |||
XM_005274651.2 | XP_005274708.1 | 2 | 87 | |||
XM_006724809.3 | XP_006724872.1 | 2 | 296 | |||
XM_011531107.2 | XP_011529409.2 | 2 | 175 | |||
XM_011531108.2 | XP_011529410.1 | 2 | 140 | |||
XM_011531110.2 | XP_011529412.1 | 2 | 192 | |||
XM_011531111.2 | XP_011529413.1 | 2 | 110 | |||
XM_011531112.2 | XP_011529414.1 | 2 | 83 | |||
XM_017029277.1 | XP_016884766.1 | 2 | 395 | |||
XM_017029278.1 | XP_016884767.1 | 3 | 299 | |||
XM_017029279.1 | XP_016884768.1 | 2 | 56 | |||
XM_017029280.1 | XP_016884769.1 | 2 | 87 | |||
GenBank mRNA | AK296067.1 | - | 2 | 80 | ||
AK296165.1 | - | 2 | 325 | |||
AK296203.1 | - | 2 | 470 | |||
AK303746.1 | - | 2 | 104 | |||
AK309159.1 | - | 1 | 35 | |||
AK316402.1 | - | 2 | 329 | |||
BC064422.1 | - | 2 | 55 | 5751849 | ||
DC383841.1 | - | 2 | 464 |