Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039582.3 | NP_001034671.3 | 6 | 898 | ||
NM_001135740.1 | NP_001129212.1 | 6 | 532 | |||
XM_005274651.2 | XP_005274708.1 | 6 | 515 | |||
XM_006724809.3 | XP_006724872.1 | 6 | 724 | |||
XM_011531107.2 | XP_011529409.2 | 6 | 603 | |||
XM_011531108.2 | XP_011529410.1 | 6 | 568 | |||
XM_011531110.2 | XP_011529412.1 | 6 | 620 | |||
XM_011531111.2 | XP_011529413.1 | 6 | 538 | |||
XM_011531112.2 | XP_011529414.1 | 6 | 511 | |||
XM_017029277.1 | XP_016884766.1 | 6 | 823 | |||
XM_017029278.1 | XP_016884767.1 | 7 | 727 | |||
XM_017029279.1 | XP_016884768.1 | 6 | 484 | |||
XM_017029280.1 | XP_016884769.1 | 6 | 515 | |||
GenBank mRNA | AK296067.1 | - | 5 | 819 | ||
AK296165.1 | - | 6 | 753 | |||
AK296203.1 | - | 6 | 898 | |||
AK303746.1 | - | 6 | 532 | |||
AK309148.1 | - | 4 | 1161 | |||
AK309159.1 | - | 2 | 645 | |||
AK316402.1 | - | 5 | 1068 | |||
BC064422.1 | - | 5 | 590 | 5751849 |