Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080473.4 | NP_536721.1 | 7 | 1142 | ||
XM_006723699.3 | XP_006723762.1 | 7 | 1162 | |||
GenBank mRNA | BC047790.1 | - | 7 | 1142 | ||
BC117356.1 | - | 6 | 1105 | 40125907 | ||
BC117358.1 | - | 6 | 1105 | 40125909 |