Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRI3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080626.5 | NP_542193.3 | 2 | 389 | ||
XM_011537940.2 | XP_011536242.1 | 2 | 377 | |||
GenBank mRNA | AF284094.1 | - | 2 | 333 | ||
BC034525.1 | - | 2 | 366 | 4778946 | ||
CA488612.1 | - | 400 |