Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRI3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080626.5 | NP_542193.3 | 3 | 656 | ||
GenBank mRNA | AF283671.2 | - | 3 | 395 | ||
AF284094.1 | - | 3 | 600 | |||
AF441865.1 | - | 2 | 260 | |||
AK055703.1 | - | 1 | 177 | |||
BC034525.1 | - | 3 | 633 | 4778946 |