Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 18 | 2142 | ||
NM_001320421.1 | NP_001307350.1 | 19 | 2189 | |||
XM_005267367.1 | XP_005267424.1 | 16 | 1666 | |||
XM_011536497.1 | XP_011534799.1 | 20 | 2413 | |||
XM_017021044.1 | XP_016876533.1 | 20 | 2319 | |||
XM_017021045.1 | XP_016876534.1 | 19 | 2157 | |||
XM_017021046.1 | XP_016876535.1 | 18 | 2200 | |||
XM_017021047.1 | XP_016876536.1 | 17 | 1817 | |||
GenBank mRNA | AK126571.1 | - | 9 | 875 | ||
AL832848.1 | - | 17 | 1820 | |||
BC035865.2 | - | 13 | 1438 | |||
BC048270.2 | - | 12 | 1243 | |||
BX247966.1 | - | 5 | 481 | |||
BX248275.1 | - | 14 | 1434 |