Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205280.1 | NP_001192209.1 | 2 | 306 | ||
XM_017026317.1 | XP_016881806.1 | 2 | 480 | |||
XM_017026318.1 | XP_016881807.1 | 2 | 1359 | |||
XM_017026319.1 | XP_016881808.1 | 2 | 480 | |||
XM_017026320.1 | XP_016881809.1 | 3 | 561 | |||
XM_017026321.1 | XP_016881810.1 | 3 | 588 | |||
XM_017026322.1 | XP_016881811.1 | 3 | 867 | |||
XM_017026323.1 | XP_016881812.1 | 2 | 480 | |||
XM_017026324.1 | XP_016881813.1 | 2 | 480 | |||
XM_017026325.1 | XP_016881814.1 | 2 | 480 | |||
GenBank mRNA | BC127928.1 | - | 1 | 126 | ||
BC127929.1 | - | 1 | 126 | |||
BX112065.1 | - | 2 | 306 |