Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541795.2 | XP_011540097.1 | 7 | 1192 | ||
XM_017001812.1 | XP_016857301.1 | 4 | 1799 | |||
XM_017001813.1 | XP_016857302.1 | 4 | 1799 | |||
XM_017001817.1 | XP_016857306.1 | 6 | 1117 | |||
XM_017001818.1 | XP_016857307.1 | 7 | 1015 | |||
XM_017001820.1 | XP_016857309.1 | 7 | 1015 | |||
XM_017001823.1 | XP_016857312.1 | 7 | 1015 | |||
XM_017001826.1 | XP_016857315.1 | 5 | 1488 | |||
XM_017001827.1 | XP_016857316.1 | 6 | 1386 | |||
GenBank mRNA | AK075260.1 | - | 2 | 1599 |
Gene Symbol: DCDC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099434.1 | NP_001092904.1 | 9 | 968 | ||
GenBank mRNA | BC041458.1 | - | 7 | 1486 | ||
BC128073.1 | - | 9 | 968 | 40109068 | ||
BM980133.1 | - | 1 | 396 |