Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100874.2 | NP_001094344.1 | 3 | 327 | ||
NM_139173.3 | NP_631912.2 | 3 | 327 | |||
NR_047513.1 | 3 | 327 | ||||
NR_047515.1 | 3 | 327 | ||||
XM_006714093.3 | XP_006714156.1 | 4 | 346 | |||
XM_011531622.1 | XP_011529924.1 | 4 | 310 | |||
XM_011531623.1 | XP_011529925.1 | 3 | 314 | |||
GenBank mRNA | AF447585.1 | - | 3 | 307 | ||
AK313592.1 | - | 3 | 254 | |||
AY461581.1 | - | 3 | 275 | |||
BC022079.1 | - | 3 | 243 | |||
BC046636.1 | - | 3 | 251 | |||
BC110794.1 | - | 3 | 211 | 5745227 | ||
BC132712.1 | - | 3 | 254 | 40146734 | ||
BC136966.1 | - | 3 | 254 | 9020958 | ||
DQ003059.1 | - | 3 | 307 |