Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001588.2 | 1 | 343 | |||
NR_024109.1 | 1 | 343 | ||||
NR_024110.1 | 1 | 343 | ||||
NR_024111.1 | 1 | 343 | ||||
GenBank mRNA | BQ441749.1 | - | 1 | 202 | ||
CN267170.1 | - | 1 | 228 | |||
CX867149.1 | - | 1 | 235 | |||
DA143511.1 | - | 1 | 346 |