Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003086.1 | 5 | 586 | |||
GenBank mRNA | AF165514.1 | - | 6 | 632 | ||
AF165517.1 | - | 5 | 582 | |||
AK293968.1 | - | 5 | 579 | |||
AY829267.1 | - | 5 | 595 | |||
BC071987.1 | - | 5 | 586 |