Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008723.1 | NP_001008723.1 | 6 | 969 | ||
XM_011539377.2 | XP_011537679.1 | 6 | 1688 | |||
XM_017015787.1 | XP_016871276.1 | 6 | 1340 | |||
XM_017015788.1 | XP_016871277.1 | 6 | 1871 | |||
XM_017015789.1 | XP_016871278.1 | 6 | 958 | |||
XM_017015790.1 | XP_016871279.1 | 6 | 958 | |||
GenBank mRNA | AK093227.1 | - | 6 | 957 | ||
AK296463.1 | - | 6 | 962 | |||
BC036225.1 | - | 6 | 971 | 5272786 | ||
BC109126.2 | - | 6 | 923 | 40036085 | ||
BX641120.1 | - | 6 | 968 |