Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270614.1 | NP_001257543.1 | 4 | 676 | ||
NM_001270615.1 | NP_001257544.1 | 4 | 743 | |||
NM_133328.3 | NP_579874.1 | 4 | 758 | |||
NR_073046.1 | 3 | 392 | ||||
NR_073047.1 | 3 | 310 | ||||
NR_073048.1 | 3 | 392 | ||||
NR_073049.1 | 3 | 392 | ||||
NR_073050.1 | 3 | 377 | ||||
NR_073051.1 | 3 | 392 | ||||
XM_011526569.1 | XP_011524871.1 | 4 | 682 | |||
XM_011526571.1 | XP_011524873.1 | 4 | 682 | |||
XM_011526572.1 | XP_011524874.1 | 4 | 682 | |||
XM_017026402.1 | XP_016881891.1 | 4 | 676 | |||
XM_017026403.1 | XP_016881892.1 | 4 | 682 | |||
GenBank mRNA | AF443591.1 | - | 3 | 559 | ||
AF457575.1 | - | 3 | 559 | |||
AK075328.1 | - | 4 | 682 | |||
AK130203.1 | - | 3 | 300 | |||
AK307895.1 | - | 4 | 651 | |||
BC027930.1 | - | 4 | 627 | 5199772 | ||
BM908148.1 | - | 3 | 304 | |||
BX374729.2 | - | 4 | 643 | |||
BX458265.2 | - | 3 | 343 | |||
CN373297.1 | - | 3 | 349 | |||
CT000394.1 | - | 3 | 306 | |||
HY033141.1 | - | 3 | 313 | |||
HY033487.1 | - | 3 | 301 |