Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175747.2 | NP_786923.1 | 1 | 1027 | ||
GenBank mRNA | AK096362.1 | - | 1 | 1027 | ||
BC051352.1 | - | 1 | 1018 | 6584192 |