Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010977.2 | NP_001010977.1 | 1 | 95 | ||
XM_017020407.1 | XP_016875896.1 | 2 | 989 | |||
XM_017020408.1 | XP_016875897.1 | 2 | 1542 | |||
GenBank mRNA | BC132748.1 | - | 1 | 95 | 40146770 | |
BC132750.1 | - | 1 | 95 | 40146772 |