Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153486.3 | NP_705690.2 | 7, 8 | 1084 | ||
NM_194436.2 | NP_919417.1 | 7, 8 | 1015 | |||
GenBank mRNA | AY092767.1 | - | 7, 8 | 1020 | ||
BC040279.1 | - | 7, 8 | 1049 | 5162826 | ||
BC047902.1 | - | 7, 8 | 1079 | 5762691 |