Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBA57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010867.3 | NP_001010867.1 | 3 | 744 | ||
NM_001310327.1 | NP_001297256.1 | 3 | 455 | |||
GenBank mRNA | AK022796.1 | - | 4 | 2771 | ||
AK303837.1 | - | 3 | 455 | |||
BI820993.1 | - | 3 | 484 |