Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBATA SIRNA Assays ›
Gene Symbol:
TBATA
Gene Name:
thymus, brain and testes associated
Gene Aliases:
C10orf27, SPATIAL
Chromosome Location:
Chr.10: 70771237 - 70785419 on Build GRCh38
UniGene ID:
Hs.386698
Species:
Human
siRNA ID
HSS153557
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBATA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318241.1
NP_001305170.1
4
615
NM_001318242.1
NP_001305171.1
4
682
NM_001318243.1
NP_001305172.1
4
615
NM_152710.3
NP_689923.3
4
615
NR_134531.1
4
615
NR_134532.1
2
387
NR_134533.1
2
387
NR_134534.1
2
387
XM_017015845.1
XP_016871334.1
4
597
XM_017015846.1
XP_016871335.1
4
597
XM_017015847.1
XP_016871336.1
4
597
XM_017015848.1
XP_016871337.1
4
597
XM_017015849.1
XP_016871338.1
4
597
XM_017015850.1
XP_016871339.1
4
597
XM_017015851.1
XP_016871340.1
4
597
XM_017015852.1
XP_016871341.1
4
597
XM_017015853.1
XP_016871342.1
4
597
XM_017015854.1
XP_016871343.1
4
597
XM_017015855.1
XP_016871344.1
4
597
XM_017015856.1
XP_016871345.1
4
597
XM_017015857.1
XP_016871346.1
4
597
XM_017015858.1
XP_016871347.1
4
597
XM_017015859.1
XP_016871348.1
4
597
XM_017015860.1
XP_016871349.1
4
597
XM_017015861.1
XP_016871350.1
4
597
XM_017015862.1
XP_016871351.1
4
597
GenBank mRNA
AK057382.1
-
4
597
AK303242.1
-
1
180
AK303344.1
-
1
180
AK303491.1
-
2
410
AK310699.1
-
1
180
AK310732.1
-
1
180
BC137543.1
-
4
649
9021543
BC137544.1
-
4
583
9021544
BC139728.1
-
4
564
40036949
BC144692.1
-
4
583
9053226
BC144694.1
-
4
583
9053228
DB147708.1
-
1
180
DB147724.1
-
1
180
DC406786.1
-
2
387
Back To Top
More Information
Gene Ontology Categories:
Function(s)
multicellular organism development
spermatogenesis
cell differentiation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0