Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178507.2 | NP_848602.1 | 1, 2 | 472 | ||
GenBank mRNA | AY459296.1 | - | 1, 2 | 231 | ||
BC047726.1 | - | 1, 2 | 472 | 5763917 |