Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143944.1 | NP_001137416.1 | 4, 5 | 446 | ||
NM_181336.3 | NP_851853.1 | 5, 6 | 1027 | |||
GenBank mRNA | AK074959.1 | - | 5, 6 | 1080 | ||
AK301083.1 | - | 4, 5 | 440 | |||
AK309888.1 | - | 1, 2 | 204 | |||
BC051803.1 | - | 5, 6 | 1017 | 5240212 | ||
DC340017.1 | - | 4, 5 | 505 |