Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 3098 | ||
XM_006715022.3 | XP_006715085.1 | 5 | 3127 | |||
XM_011514394.2 | XP_011512696.1 | 5 | 3140 | |||
XM_011514395.2 | XP_011512697.1 | 5 | 4129 | |||
XM_017010478.1 | XP_016865967.1 | 5 | 3124 | |||
GenBank mRNA | BC101628.1 | - | 4 | 3057 | 8069134 | |
BC112275.1 | - | 4 | 3057 | 8327743 |