Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136482.1 | NP_001129954.1 | 2 | 391 | ||
XM_005259846.4 | XP_005259903.1 | 3 | 737 | |||
XM_005259847.4 | XP_005259904.1 | 3 | 579 | |||
GenBank mRNA | BC157863.1 | - | 2 | 391 | 9057082 |