Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190259.1 | NP_001177188.1 | 5, 6 | 398 | ||
NM_001190260.1 | NP_001177189.1 | 4, 5 | 347 | |||
NM_152785.4 | NP_689998.1 | 5, 6 | 392 | |||
XM_005247272.2 | XP_005247329.1 | 5, 6 | 816 | |||
GenBank mRNA | AF521911.1 | - | 5, 6 | 379 | ||
AK313368.1 | - | 5, 6 | 339 | |||
AY212246.1 | - | 5, 6 | 313 | |||
BC030506.1 | - | 5, 6 | 260 | 4385178 | ||
BM456595.1 | - | 5, 6 | 348 | |||
BP224611.1 | - | 4, 5 | 295 | |||
BX280641.1 | - | 5, 6 | 448 | |||
DA455714.1 | - | 5, 6 | 393 |