Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025015.2 | NP_079291.2 | 5 | 568 | ||
XM_005269672.3 | XP_005269729.1 | 6 | 718 | |||
XM_005269673.4 | XP_005269730.1 | 6 | 1662 | |||
XM_011539579.2 | XP_011537881.1 | 7 | 779 | |||
XM_011539580.2 | XP_011537882.1 | 6 | 724 | |||
GenBank mRNA | AB007877.1 | - | 5 | 568 | ||
AK294672.1 | - | 4 | 382 |