Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161812.1 | NP_001155284.1 | 2 | 219 | ||
NM_001161814.1 | NP_001155286.1 | 3 | 413 | |||
NM_172370.4 | NP_758958.3 | 2 | 173 | |||
XM_005254042.1 | XP_005254099.1 | 2 | 371 | |||
GenBank mRNA | AY138546.1 | - | 2 | 146 | ||
AY138547.1 | - | 3 | 413 | |||
AY170469.2 | - | 3 | 235 | |||
AY170470.2 | - | 3 | 235 | |||
AY223901.1 | - | 2 | 371 | |||
BC121091.1 | - | 2 | 141 | 40119921 | ||
DQ343761.1 | - | 2 | 420 | |||
DQ357223.1 | - | 2 | 165 | |||
DQ386869.1 | - | 2 | 119 | |||
DQ386870.1 | - | 2 | 165 |