Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161812.1 | NP_001155284.1 | 3 | 347 | ||
NM_001161814.1 | NP_001155286.1 | 4 | 541 | |||
NM_172370.4 | NP_758958.3 | 3 | 301 | |||
XM_005254042.1 | XP_005254099.1 | 3 | 499 | |||
GenBank mRNA | AY138546.1 | - | 3 | 274 | ||
AY138547.1 | - | 4 | 541 | |||
AY170469.2 | - | 4 | 363 | |||
AY170470.2 | - | 4 | 363 | |||
AY170471.1 | - | 3 | 241 | |||
AY223901.1 | - | 3 | 499 | |||
BC121091.1 | - | 3 | 269 | 40119921 | ||
DQ343761.1 | - | 3 | 548 | |||
DQ357223.1 | - | 3 | 293 | |||
DQ386869.1 | - | 3 | 247 | |||
DQ386870.1 | - | 3 | 293 |