Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024340.1 | 2 | 1673 | |||
GenBank mRNA | BC038741.1 | - | 2 | 1674 | ||
BC119749.2 | - | 1 | 549 | |||
BC119750.2 | - | 1 | 549 | |||
BC126326.1 | - | 1 | 884 | |||
BC130402.1 | - | 1 | 884 | |||
CR933684.1 | - | 3 | 1399 |