Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037225.2 | NP_001032302.1 | 10 | 916 | ||
NM_001300803.1 | NP_001287732.1 | 8 | 746 | |||
NM_001318808.1 | NP_001305737.1 | 11 | 1694 | |||
XM_005273918.2 | XP_005273975.1 | 11 | 974 | |||
XM_006718502.2 | XP_006718565.1 | 11 | 1091 | |||
XM_006718503.3 | XP_006718566.1 | 11 | 977 | |||
XM_006718504.2 | XP_006718567.1 | 10 | 1027 | |||
XM_006718505.2 | XP_006718568.1 | 8 | 857 | |||
XM_011544940.2 | XP_011543242.1 | 11 | 972 | |||
XM_011544941.2 | XP_011543243.1 | 5 | 673 | |||
GenBank mRNA | AK093130.1 | - | 11 | 1688 | ||
BC033501.1 | - | 8 | 678 | |||
BC106951.2 | - | 9 | 711 | 40035116 | ||
BC106952.1 | - | 9 | 711 | 40035117 | ||
LC068587.1 | - | 9 | 818 |