Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126496.1 | 1 | 823 | |||
NR_126497.1 | 1 | 967 | ||||
NR_126498.1 | 1 | 1824 | ||||
NR_126500.1 | 1 | 967 | ||||
GenBank mRNA | AK090420.1 | - | 1 | 823 | ||
AK091232.1 | - | 1 | 1823 | |||
BC128083.1 | - | 1 | 1546 | 40075904 | ||
BC144351.1 | - | 1 | 966 | 9052871 |