Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST20-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028330.1 | 1 | 49 | |||
GenBank mRNA | AK091107.1 | - | 1 | 49 | ||
BC101201.1 | - | 1 | 49 | 40021818 | ||
BC101202.1 | - | 1 | 49 | 40021819 | ||
BC101203.1 | - | 1 | 49 | 40021821 |