Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1680 | ||
NR_003494.1 | 6 | 1414 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1775 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1845 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1589 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1432 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1854 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1793 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1821 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1742 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1714 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1303 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1277 | ||
BC062615.1 | - | 7 | 1354 | |||
BC085016.1 | - | 8 | 1383 | |||
BC093808.1 | - | 1 | 229 | |||
BC093810.1 | - | 1 | 229 | |||
BC098427.1 | - | 7 | 1360 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1710 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1629 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1530 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1568 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1731 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1782 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1749 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1651 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1712 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1649 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1631 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 8 | 1772 | |||
GenBank mRNA | BC043342.2 | - | 7 | 1496 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 1277 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 1388 | |||
GenBank mRNA | AK124448.1 | - | 3 | 1156 |