Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029955.3 | NP_001025126.2 | 1 | 548 | ||
GenBank mRNA | AK122914.1 | - | 1 | 466 | ||
BC035027.1 | - | 1 | 421 | 4828469 | ||
BC052943.1 | - | 1 | 488 | 5268527 | ||
BC104224.1 | - | 1 | 470 | 40032133 | ||
BC104225.1 | - | 1 | 470 | 40032134 | ||
DB100231.1 | - | 1 | 548 |