Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005783.3 | NP_005774.2 | 4 | 661 | ||
XM_017003147.1 | XP_016858636.1 | 4 | 620 | |||
GenBank mRNA | AB006679.1 | - | 4 | 599 | ||
AB451403.1 | - | 3 | 470 | |||
AK301722.1 | - | 5 | 631 | |||
AK313777.1 | - | 4 | 581 | |||
BC005968.1 | - | 4 | 597 | 4073821 | ||
BC022864.1 | - | 4 | 601 | 4096526 | ||
BC024223.2 | - | 4 | 601 | 4107372 | ||
BC070183.1 | - | 4 | 595 | 4551680 | ||
CR456935.1 | - | 3 | 470 |